missing translation for 'onlineSavingsMsg'
Learn More

Trimethylacetic anhydride, 99%, Thermo Scientific™

Catalog No. AC162440010
Change view
Click to view available options
1L, Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC162440010 1L, Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC162440010 Supplier Thermo Scientific Chemicals Supplier No. 162440010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1538-75-6
Density 0.9100g/mL
Boiling Point 193.0°C
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H18O3
Refractive Index 1.4080 to 1.4100
Linear Formula [(CH3)3CCO]2O
Quantity 1L
MDL Number MFCD00008842
Beilstein 02,32
Specific Gravity 0.91
Synonym pivalic anhydride, trimethylacetic anhydride, 2,2-dimethylpropionic anhydride, propanoic acid, 2,2-dimethyl-, anhydride, pivalic anydride, 2,2-dimethylpropanoic anhydride, trimethylacetic acid anhydride, propanoic acid, 2,2-dimethyl-, 1,1'-anhydride, trimethylaceticanhydride, bispivalic anhydride
Solubility Information Solubility in water: decomposes in water
InChI Key PGZVFRAEAAXREB-UHFFFAOYSA-N
SMILES CC(C)(C)C(=O)OC(=O)C(C)(C)C
IUPAC Name 2,2-dimethylpropanoyl 2,2-dimethylpropanoate
Molecular Weight (g/mol) 186.25
PubChem CID 15234
Formula Weight 186.25
Percent Purity 99%
Chemical Name or Material Trimethylacetic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 216-263-1

RUO – Research Use Only