missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Phosphate 98.0+%, TCI America™

Catalog No. P0271500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0271500G 500 g
P027125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0271500G Supplier TCI America Supplier No. P0271500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 512-56-1
Molecular Formula C3H9O4P
Molecular Weight (g/mol) 140.08
MDL Number MFCD00008348
InChI Key WVLBCYQITXONBZ-UHFFFAOYSA-N
Synonym trimethylphosphate, phosphoric acid, trimethyl ester, trimethyl orthophosphate, tmpo, trimethoxyphosphine oxide, phosphoric acid trimethyl ester, o,o,o-trimethyl phosphate, trimethylfosfat, phosphate, trimethyl, trimethylfosfat czech
PubChem CID 10541
ChEBI CHEBI:46324
IUPAC Name trimethyl phosphate
SMILES COP(=O)(OC)OC

Specifications

Melting Point -46°C
Color Colorless
Boiling Point 197°C
Quantity 500 g
Formula Weight 140.07
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Phosphate
EINECSNumber (2)-2000
RTECSNumber TC8225000
TSCA Yes