missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethyl(phenoxy)silane 97.0+%, TCI America™
Supplier: TCI America T322225ML
Specifications
| Trimethyl(phenoxy)silane | |
| -55°C | |
| C9H14OSi | |
| 25 mL | |
| trimethyl phenoxy silane, phenoxytrimethylsilane, silane, trimethylphenoxy, phenyl trimethylsilyl ether, trimethylsilylphenoxide, trimethylsiloxy benzene, phenol, tms ether, phenol trimethylsilyl ether, benzene, trimethylsilyl oxy | |
| C[Si](C)(C)OC1=CC=CC=C1 | |
| 166.30 | |
| 166.30 | |
| Liquid |
| 1529-17-5 | |
| 211°C | |
| MFCD00053666 | |
| 1993 | |
| OJAJJFGMKAZGRZ-UHFFFAOYSA-N | |
| trimethyl(phenoxy)silane | |
| 73720 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1529-17-5 | |
| 166.30 | |
| OJAJJFGMKAZGRZ-UHFFFAOYSA-N | |
| 73720 | |
| C[Si](C)(C)OC1=CC=CC=C1 |
| C9H14OSi | |
| MFCD00053666 | |
| trimethyl phenoxy silane, phenoxytrimethylsilane, silane, trimethylphenoxy, phenyl trimethylsilyl ether, trimethylsilylphenoxide, trimethylsiloxy benzene, phenol, tms ether, phenol trimethylsilyl ether, benzene, trimethylsilyl oxy | |
| trimethyl(phenoxy)silane |
Safety and Handling
EINECSNumber : (3)-2640
TSCA : Yes