missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(phenoxy)silane 97.0+%, TCI America™

Catalog No. p-7137645
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32225ML 5 mL
T322225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T32225ML Supplier TCI America Supplier No. T32225ML
Only null left

Chemical Identifiers

CAS 1529-17-5
Molecular Formula C9H14OSi
Molecular Weight (g/mol) 166.30
MDL Number MFCD00053666
InChI Key OJAJJFGMKAZGRZ-UHFFFAOYSA-N
Synonym trimethyl phenoxy silane, phenoxytrimethylsilane, silane, trimethylphenoxy, phenyl trimethylsilyl ether, trimethylsilylphenoxide, trimethylsiloxy benzene, phenol, tms ether, phenol trimethylsilyl ether, benzene, trimethylsilyl oxy
PubChem CID 73720
IUPAC Name trimethyl(phenoxy)silane
SMILES C[Si](C)(C)OC1=CC=CC=C1

Specifications

Melting Point -55°C
Boiling Point 211°C
Quantity 5 mL
UN Number 1993
Formula Weight 166.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl(phenoxy)silane
EINECSNumber (3)-2640
TSCA Yes