CAS RN 1529-17-5
CAS RN 1529-17-5
IUPAC Name:
trimethyl(phenoxy)silane
Synonyms:
silane, trimethylphenoxy-
Molecular Weight (g/mol):
166.3
Molecular Formula:
C9H14OSi
InChi Key:
OJAJJFGMKAZGRZ-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OC1=CC=CC=C1
1
–
1
of
1
results
1
Trimethyl(phenoxy)silane 97.0+%, TCI America™
CAS: 1529-17-5 Molecular Formula: C9H14OSi Molecular Weight (g/mol): 166.30 MDL Number: MFCD00053666 InChI Key: OJAJJFGMKAZGRZ-UHFFFAOYSA-N Synonym: trimethyl phenoxy silane,phenoxytrimethylsilane,silane, trimethylphenoxy,phenyl trimethylsilyl ether,trimethylsilylphenoxide,trimethylsiloxy benzene,phenol, tms ether,phenol trimethylsilyl ether,benzene, trimethylsilyl oxy PubChem CID: 73720 IUPAC Name: trimethyl(phenoxy)silane SMILES: C[Si](C)(C)OC1=CC=CC=C1