missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Orthopropionate 97.0+%, TCI America™

Numéro de catalogue. T22885G
Modifier l'affichage
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T22885G 5 g
T228825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T22885G Fournisseur TCI America Code fournisseur T22885G
Il en reste null

Identifiants chimiques

CAS 24823-81-2
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00043305
InChI Key ZGMNAIODRDOMEK-UHFFFAOYSA-N
Synonym trimethyl orthopropionate, propane, 1,1,1-trimethoxy, trimethylorthopropionate, trimethoxypropane, acmc-209gew, trimethyl orthopropanoate, propane,1,1,1-trimethoxy, ksc489s6b, zgmnaiodrdomek-uhfffaoysa, orthopropionic acid trimethyl ester
PubChem CID 141145
IUPAC Name 1,1,1-trimethoxypropane
SMILES CCC(OC)(OC)OC

Spécifications

Color Colorless
Boiling Point 128°C
Quantity 5 g
UN Number 3272
Formula Weight 134.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl Orthopropionate