missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl(methylcyclopentadienyl)platinum(IV)

Catalog No. AA9815802
Change view
Click to view available options
Quantity:
0.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA9815802 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA9815802 Supplier Thermo Scientific Chemicals Supplier No. 09815802

CAS: 94442-22-5 | C9H16Pt | 319.31 g/mol

Trimethyl(methylcyclopentadienyl)platinum(IV) is used as an organometallic precursor for platinum. It is used in the microelectronics, catalysis, photonics and chemical sensing processes. It is used for the deposition of ultrathin metallic layers of platinum by the electron beam induced deposition process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94442-22-5
Molecular Formula C9H16Pt
Molecular Weight (g/mol) 319.31
MDL Number MFCD00079665
InChI Key AGQKROMSWCHOND-UHFFFAOYSA-N
IUPAC Name trimethyl(2-methylcyclopenta-1,3-dien-1-yl)platinum
SMILES C[Pt](C)C.Cc1cccc1

Specifications

Melting Point 28°C to 31°C
Color White
Boiling Point 23°C (0.053 mmHg)(sublimation)
Quantity 0.5 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 319.32
Physical Form Pieces or Powder
Chemical Name or Material Trimethyl (methylcyclopentadienyl)platinum (IV)
Hazard Category H301+H311
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only