missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl Borate 98.0+%, TCI America™

Numéro de catalogue. p-7118496
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B052225ML 25 mL
B0522100ML 100 mL
B0522500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. B052225ML Fournisseur TCI America Code fournisseur B052225ML
Il en reste null

Identifiants chimiques

CAS 121-43-7
Molecular Formula C3H9BO3
Molecular Weight (g/mol) 103.912
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Spécifications

Melting Point -29°C
Color Yellow
Boiling Point 68°C
Quantity 25 mL
UN Number 2416
Formula Weight 103.91
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trimethyl Borate
EINECSNumber (2)-2101&(2)-2114
RTECSNumber ED5600000
TSCA Yes