missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl borate, 99%, Thermo Scientific Chemicals

Catalog No. AAB20215AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB20215AU 1000 mL
AAB20215AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB20215AU Supplier Thermo Scientific Chemicals Supplier No. B20215AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-43-7 | Molecular Formula: C3H9BO3 | Molecular Weight: 103.912 g/mol

Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trimethyl borate is a useful reagent in organic synthesis. It is involved in the production of resins, waxes and paints and acts as a methylation agent. As a boron source, it is used to prepare flame retardants, anti-oxidants and corrosion inhibitors. It reacts with Grignard reagents followed by hydrolysis to prepare boronic acid. It is also used as a precursor of borate esters, which finds application in Suzuki coupling reaction. It is an intermediate in the preparation of sodium borohydride.

Solubility
Miscible with tetrahydrofuran, ether, isoporpylamine, hexane and methanol.

Notes
Moisture sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It decomposes slowly in the presence of moisture and rapidly in water.

Chemical Identifiers

CAS 121-43-7
Molecular Formula C3H9BO3
Molecular Weight (g/mol) 103.912
MDL Number MFCD00008346
InChI Key WRECIMRULFAWHA-UHFFFAOYSA-N
Synonym trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
PubChem CID 8470
ChEBI CHEBI:38913
IUPAC Name trimethyl borate
SMILES B(OC)(OC)OC

Specifications

Melting Point -34°C
Density 0.915
Boiling Point 67°C to 69°C
Flash Point −8°C (17°F)
Odor Odorless
Refractive Index 1.361
Linear Formula (CH3O)3B
Quantity 1000 mL
UN Number UN2416
Beilstein 1697939
Sensitivity Moisture sensitive
Merck Index 14,9712
Solubility Information Miscible with tetrahydrofuran,ether,isoporpylamine,hexane and methanol.
Formula Weight 103.92
Percent Purity 99%
Chemical Name or Material Trimethyl borate
Show More Show Less
Hazard Category H225-H312-H370
Hazard Statement GHS H Statement
H225-H312
Highly flammable liquid and vapour.
Harmful in contact with skin.
Precautionary Statement P210-P233-P240-P243-P260-P264b-P270-P280-P302+P352-P312-P314-P362-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIMETHYL BORATE
EINECSNumber 204-468-9
RTECSNumber ED5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only