missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl 1,2,4-Cyclohexanetricarboxylate (cis- and trans- mixture) 97.0+%, TCI America™

Catalog No. T30295G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30295G 5 g
T30291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30295G Supplier TCI America Supplier No. T30295G

Chemical Identifiers

CAS 185855-30-5
Molecular Formula C12H18O6
Molecular Weight (g/mol) 258.27
InChI Key DEIJHCATUHBNIH-UHFFFAOYSA-N
Synonym 1,2,4-Cyclohexanetricarboxylic Acid Trimethyl Ester
PubChem CID 12616206
IUPAC Name trimethyl cyclohexane-1,2,4-tricarboxylate
SMILES COC(=O)C1CCC(C(C1)C(=O)OC)C(=O)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 258.27
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Trimethyl 1,2,4-Cyclohexanetricarboxylate (cis- and trans- mixture)
TSCA No
Recommended Storage Refrigerator