missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRIM65 Monoclonal Antibody (K52045_2F4), Invitrogen™

Numéro de catalogue. MA555752
Modifier l'affichage
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
MA555752 50 μL
MA556132 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. MA555752 Fournisseur Invitrogen™ Code fournisseur MA555752
Il en reste null

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DAERLKREAQ LRASLEVTQQ QATQAEGQLL ELRKQSSQIQ NSACILASWV SGKFSSLLQA LEIQHTTALR SIEVAKTQAL AQARDEEQRL RVHLEAVARH GCRIRELLEQ VDEQTFLQES QLLQPPGPLG PLTPLQWDED QQLGDLKQLL SRLCGLLLEE GSHPGAPAKP VDLAPVEAPG

TRIM65 is a protein coding gene and gene ontology (GO) includes cytosol, nucleoplasm, nucleus, positive regulation of autophagy.

Spécifications

Antigen TRIM65
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K52045_2F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TRIM65
Gene Accession No. Q6PJ69
Gene Alias 4732463G12Rik; TRIM65; tripartite motif containing 65; tripartite motif-containing protein 65
Gene Symbols TRIM65
Host Species Mouse
Immunogen Recombinant human TRIM65 (immunogen range is 131-310).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201292
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Afficher plus de résultats Afficher moins de résultats