missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM47/GOA Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Gene overexpressed in astrocytoma
Spécifications
Spécifications
| Antigène | TRIM47/GOA |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/ml |
| Conjugué | Unconjugated |
| Formule | TBS with 0.1% BSA and 0.09% sodium azide |
| Gène | TRIM47 |
| Numéro d’acquisition du gène | Q96LD4 |
| Alias du gène | GOA, RNF100, RING finger protein 100, gene overexpressed in astrocytoma protein, tripartite motif-containing 47, tripartite motif-containing protein 47 |
| Symboles de gène | TRIM47 |
| Afficher plus de résultats |