missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trilinolein 95.0+%, TCI America™
Specifications
Specifications
| Quantity | 5 g |
| Formula Weight | 879.41 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Trilinolein |
Safety and Handling
| TSCA | No |
| Recommended Storage | Refrigerator |