missing translation for 'onlineSavingsMsg'
Learn More

Trilauryl Trithiophosphite, TCI America™

Catalog No. T046025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T046025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T046025G Supplier TCI America Supplier No. T046025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1656-63-9
Molecular Formula C36H75PS3
Molecular Weight (g/mol) 635.15
MDL Number MFCD00015072
InChI Key JZNDMMGBXUYFNQ-UHFFFAOYSA-N
Synonym Tridodecyl Phosphonotrithioite, Tridodecyl Trithiophosphite, Phosphorotrithious Acid Tridodecyl Ester
PubChem CID 74256
IUPAC Name tris(dodecylsulfanyl)phosphane
SMILES CCCCCCCCCCCCSP(SCCCCCCCCCCCC)SCCCCCCCCCCCC

Specifications

Color White
Quantity 25 g
Formula Weight 635.15
Physical Form Crystalline Lumps
Chemical Name or Material Trilauryl Trithiophosphite
EINECSNumber (2)-1957
TSCA Yes