missing translation for 'onlineSavingsMsg'
Learn More

Trilaurin

Catalog No. AAJ6296214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6296214 25 g
AAJ6296206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6296214 Supplier Thermo Scientific Chemicals Supplier No. J6296214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 538-24-9 | C39H74O6 | 639.015 g/mol

Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trilaurin is used in makeup products, creams and lotions, deodorants, suntan and sunscreen products, hair conditioners, and skin care and skin cleansing products. These are used in cosmetic products as thickening agents and emollients. It is used as a viscosity controlling agent.

Solubility
Soluble in water, ethylacetate- almost transparency.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 538-24-9
Molecular Formula C39H74O6
Molecular Weight (g/mol) 639.015
MDL Number MFCD00026559
InChI Key VMPHSYLJUKZBJJ-UHFFFAOYSA-N
Synonym trilaurin, glycerol trilaurate, glyceryl tridodecanoate, glyceryl trilaurate, glycerin trilaurate, lauric acid triglyceride, tridodecanoin, lauric acid triglycerin ester, trilauroylglycerol, laurin, tri
PubChem CID 10851
ChEBI CHEBI:77389
IUPAC Name 2,3-di(dodecanoyloxy)propyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCC)OC(=O)CCCCCCCCCCC

Specifications

Melting Point 47°C
Color White
Linear Formula [CH3(CH2)10COOCH2]2CHOCO(CH2)10CH3
Quantity 25 g
Beilstein 1730452
Solubility Information Soluble in water,ethylacetate- almost transparency.
Formula Weight 639.01
Physical Form Powder
Chemical Name or Material Trilaurin
EINECSNumber 208-687-0
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only