missing translation for 'onlineSavingsMsg'
Learn More

Triisopropylsilylacetylene 95.0+%, TCI America™

Catalog No. T16835ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16835ML 5 mL
T168325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16835ML Supplier TCI America Supplier No. T16835ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89343-06-6
Molecular Formula C11H22Si
Molecular Weight (g/mol) 182.382
MDL Number MFCD00075452
InChI Key KZGWPHUWNWRTEP-UHFFFAOYSA-N
Synonym triisopropylsilyl acetylene, ethynyltriisopropylsilane, triisopropylsilylacetylene, ethynyltris propan-2-yl silane, ethynyl-tri propan-2-yl silane, silane, ethynyltris 1-methylethyl, triisopropylsilylacetylen, acmc-209qzk, ethynyltriisopropyl-silane
PubChem CID 2734682
IUPAC Name ethynyl-tri(propan-2-yl)silane
SMILES CC(C)[Si](C#C)(C(C)C)C(C)C

Specifications

Color Yellow
Boiling Point 100°C
Quantity 5 mL
UN Number 1993
Formula Weight 182.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triisopropylsilylacetylene