missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisopropanolamine cyclic borate, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 150°C to 157°C |
| Color | White to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% min. (GC) |
| Packaging | Glass bottle |
| Quantity | 500 g |
| Formula Weight | 199.06 |
| Percent Purity | 98+% |
| Physical Form | Crystalline Chunks and Needles |
| Chemical Name or Material | Triisopropanolamine cyclic borate |
RUO – Research Use Only