missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine Borate 98.0+%, TCI America™

Catalog No. T29435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29435G 5 g
T294325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29435G Supplier TCI America Supplier No. T29435G
Only null left

Chemical Identifiers

CAS 101-00-8
Molecular Formula C9H18BNO3
Molecular Weight (g/mol) 199.057
MDL Number MFCD03791298
InChI Key IWKGJTDSJPLUCE-UHFFFAOYSA-N
Synonym triisopropanolamine cyclic borate, 3,7,10-trimethyl-2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, borester 21, triisopropanolamine borate, boric acid, tris 1-amino-2-propyl ester, 2-propanol, 1,1',1-nitrilotri-, cyclic borate, 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo 3.3.3 undecane, 2,8,9-trioxa-5-aza-1-borabicyclo 3.3.3 undecane, 3,7,10-trimethyl, triisopropanolamineborate
PubChem CID 225550
IUPAC Name 3,7,10-trimethyl-4,6,11-trioxa-1-aza-5-borabicyclo[3.3.3]undecane
SMILES B12OC(CN(CC(O1)C)CC(O2)C)C

Specifications

Melting Point 155°C
Color White-Yellow
Boiling Point 135°C
Quantity 5 g
Formula Weight 199.06
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Triisopropanolamine Borate
RTECSNumber ED5787000
TSCA No