missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triisobutylamine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 183°C |
| Quantity | 500 mL |
| UN Number | 2734 |
| Formula Weight | 185.36 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | Triisobutylamine |
Safety and Handling
| TSCA | Yes |