missing translation for 'onlineSavingsMsg'
Learn More

Trihexyl Borate 98.0+%, TCI America™

Catalog No. B20945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20945G 5 g
B209425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20945G Supplier TCI America Supplier No. B20945G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5337-36-0
Molecular Formula C18H39BO3
Molecular Weight (g/mol) 314.317
MDL Number MFCD00027295
InChI Key KDQYHGMMZKMQAA-UHFFFAOYSA-N
Synonym Boric Acid Trihexyl Ester
PubChem CID 21423
IUPAC Name trihexyl borate
SMILES B(OCCCCCC)(OCCCCCC)OCCCCCC

Specifications

Color Yellow
Boiling Point 311°C
Quantity 5 g
Formula Weight 314.32
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trihexyl Borate
RTECSNumber ED5120000
TSCA Yes