missing translation for 'onlineSavingsMsg'
Learn More

Trigonelline hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111014895
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014895 10 g
111014893 1 g
111014894 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111014895 Supplier Merck Emd Millipore Supplier No. T550910G
Featured Industries: Food and Beverages

Specifications

CAS 6138-41-6
Physical Form Neat
Linear Formula C7H7 NO2 · HCl
MDL Number MFCD00077250
Synonym 1-Methylpyridinium-3-carboxylate hydrochloride; N-Methylnicotinic acid betaine hydrochloride
Type Analytical Standard
Quantity 10 g
Formula Weight 173.60
Grade Analytical Standard