missing translation for 'onlineSavingsMsg'
Learn More

Triflusal 98.0+%, TCI America™

Supplier:  TCI America T36011G

 View more versions of this product

Catalog No. T36011G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Triflusal
115°C
C10H7F3O4
1 g
RMWVZGDJPAKBDE-UHFFFAOYSA-N
2-acetyloxy-4-(trifluoromethyl)benzoic acid
9458
≥98.0% (T)
322-79-2
White
MFCD00866793
Acetyl-4-(trifluoromethyl)salicylic Acid, 2-Acetoxy-4-(trifluoromethyl)benzoic Acid, Drisgen
CC(=O)OC1=C(C=CC(=C1)C(F)(F)F)C(=O)O
248.157
248.16
Crystalline Powder

Chemical Identifiers

322-79-2
248.157
RMWVZGDJPAKBDE-UHFFFAOYSA-N
9458
CC(=O)OC1=C(C=CC(=C1)C(F)(F)F)C(=O)O
C10H7F3O4
MFCD00866793
Acetyl-4-(trifluoromethyl)salicylic Acid, 2-Acetoxy-4-(trifluoromethyl)benzoic Acid, Drisgen
2-acetyloxy-4-(trifluoromethyl)benzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions