missing translation for 'onlineSavingsMsg'
Learn More

Triflusal 98.0+%, TCI America™

Catalog No. p-7137813
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3601250MG 250 mg
T36011G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3601250MG Supplier TCI America Supplier No. T3601250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 322-79-2
Molecular Formula C10H7F3O4
Molecular Weight (g/mol) 248.157
MDL Number MFCD00866793
InChI Key RMWVZGDJPAKBDE-UHFFFAOYSA-N
Synonym Acetyl-4-(trifluoromethyl)salicylic Acid, 2-Acetoxy-4-(trifluoromethyl)benzoic Acid, Drisgen
PubChem CID 9458
IUPAC Name 2-acetyloxy-4-(trifluoromethyl)benzoic acid
SMILES CC(=O)OC1=C(C=CC(=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 115°C
Color White
Quantity 250 mg
Formula Weight 248.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triflusal
TSCA No
Recommended Storage Refrigerator