accessibility menu, dialog, popup

UserName

CAS RN 322-79-2

IUPAC Name: 2-(acetyloxy)-4-(trifluoromethyl)benzoic acid
Synonyms: triflusal disgren triflusalum hormona brand of triflusal uriach brand of triflusal
Molecular Weight (g/mol): 248.16
Molecular Formula: C10H7F3O4
InChi Key: RMWVZGDJPAKBDE-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC(=CC=C1C(O)=O)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results