missing translation for 'onlineSavingsMsg'
Learn More

Trifluorothymidine 98.0+%, TCI America™

Catalog No. T25111G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25111G 1 g
T2511100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25111G Supplier TCI America Supplier No. T25111G
Only null left

Chemical Identifiers

CAS 70-00-8
Molecular Formula C10H11F3N2O5
Molecular Weight (g/mol) 296.20
MDL Number MFCD00006534
InChI Key VSQQQLOSPVPRAZ-RRKCRQDMSA-N
Synonym trifluridine, trifluorothymidine, viroptic, 5-trifluorothymidine, trifluridina, trifluridinum, virophta, trifluoromethyldeoxyuridine, f3tdr, f3dthd
PubChem CID 6256
ChEBI CHEBI:75179
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(C(=O)NC1=O)C(F)(F)F

Specifications

Melting Point 189°C
Color White
Quantity 1 g
Formula Weight 296.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trifluorothymidine
RTECSNumber XP2087500
TSCA No