missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic Anhydride 98.0+%, TCI America™

Catalog No. T110010G
Change view
Click to view available options
Quantity:
10 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T110010G 10 g
T110025G 25 g
T1100250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T110010G Supplier TCI America Supplier No. T110010G

Chemical Identifiers

CAS 358-23-6
Molecular Formula C2F6O5S2
Molecular Weight (g/mol) 282.127
MDL Number MFCD00000408
InChI Key WJKHJLXJJJATHN-UHFFFAOYSA-N
Synonym trifluoromethanesulfonic anhydride, triflic anhydride, trifluoromethanesulphonic anhydride, trifluoromethanesulfonic acid anhydride, methanesulfonic acid, trifluoro-, anhydride, trifluoromethanesulphonic acid anhydride, unii-8w034lhg1u, tf2o, trifluoromethane sulfonyl trifluoromethanesulfonate, trifluoromethane sulfonic anhydride
PubChem CID 67749
ChEBI CHEBI:48509
IUPAC Name trifluoromethylsulfonyl trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point -80°C
Color Colorless
Boiling Point 84°C
Quantity 10 g
UN Number 3265
Formula Weight 282.13
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Trifluoromethanesulfonic Anhydride
EINECSNumber (2)-3807
TSCA Yes
Recommended Storage Refrigerator