missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonamide 98.0+%, TCI America™

Catalog No. T129025G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T129025G 25 g
T129010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T129025G Supplier TCI America Supplier No. T129025G

Chemical Identifiers

CAS 421-85-2
Molecular Formula CH2F3NO2S
Molecular Weight (g/mol) 149.09
MDL Number MFCD00068714
InChI Key KAKQVSNHTBLJCH-UHFFFAOYSA-N
Synonym trifluoromethanesulphonamide, trifluoromethane sulfonamide, trifluoromethylsulfonamide, 1,1,1-trifluoromethanesulfonamide, chembl67511, methanesulfonamide, 1,1,1-trifluoro, 1bcd, trifluoromethyl sulfonamide, triflamide, trifluorometanesulfonamide
PubChem CID 79001
IUPAC Name trifluoromethanesulfonamide
SMILES NS(=O)(=O)C(F)(F)F

Specifications

Melting Point 120°C
Color White
Quantity 25 g
Formula Weight 149.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trifluoromethanesulfonamide