missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic anhydride, TFAA, EMD-Millipore

Numéro de catalogue. m1125130010
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M1125130010 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M1125130010 Fournisseur Technidata Code fournisseur 1.12513.0010
Il en reste null

Identifiants chimiques

CAS 407-25-0
Molecular Formula C4F6O3
Molecular Weight (g/mol) 210.03
MDL Number MFCD00000416
InChI Key QAEDZJGFFMLHHQ-UHFFFAOYSA-N
Synonym Bis(trifluoroacetic) anhydride, TFAA
PubChem CID 9845
IUPAC Name 2,2,2-trifluoroacetyl 2,2,2-trifluoroacetate
SMILES FC(F)(F)C(=O)OC(=O)C(F)(F)F

Spécifications

Packaging Glass
Vapor Pressure 405 hPa (20°C)
Density Density: 1.51g/cm3 (20°C)
Linear Formula (CF3CO)2O
Quantity 10 mL
Formula Weight 210.03
Chemical Name or Material Trifluoroacetic anhydride
Recommended Storage Store at 15° to 25°C