missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid 99.0+%, TCI America™
Specifications
Specifications
| Boiling Point | 73°C |
| Color | Colorless |
| Quantity | 500 g |
| Physical Form | Liquid |
| UN Number | 2699 |
| Formula Weight | 114.02 |
| Percent Purity | ≥99.0% (T) |
| Chemical Name or Material | Trifluoroacetic Acid |
Safety and Handling
| EINECSNumber | (2)-1185 |
| RTECSNumber | AJ9625000 |
| TSCA | Yes |