missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Solstice
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Density | 1.489g/mL at 20°C |
| Quantity | 5 x 10 Ampules |
| Flash Point | >100°C (>212°F) |
| Linear Formula | CF3COOH |
| Vapor Density | 3.9 (vs air) |
| UN Number | UN2699 |
| Vapor Pressure | 97.5 mmHg (20°C) |
| Formula Weight | 114.02 |
| Percent Purity | 99% |
| Chemical Name or Material | Trifluoroacetic acid |
Safety and Handling
| Hazard Statement | P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| DOTInformation | 8 / PGI |
| RTECSNumber | 200-929-3 |