missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™
Reagent Grade, 99%
Supplier: Honeywell Inc T6508500ML
Description
- ≥95% for general laboratory use.
Specifications
| Trifluoroacetic acid | |
| C2HF3O2 | |
| MFCD00004169 | |
| 742035 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| trifluoroacetic acid | |
| 6422 | |
| 114.02g/mol | |
| ReagentPlus | |
| 500 mL | |
| 3.9 (vs air) |
| 76-05-1 | |
| CF3COOH | |
| UN2699 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| OC(=O)C(F)(F)F | |
| 114.02 | |
| CHEBI:45892 | |
| 99% | |
| 1.489g/mL at 20°C | |
| >100°C (>212°F) | |
| 97.5 mmHg (20°C) |
Chemical Identifiers
| 76-05-1 | |
| 114.02 | |
| DTQVDTLACAAQTR-UHFFFAOYSA-N | |
| 6422 | |
| trifluoroacetic acid |
| C2HF3O2 | |
| MFCD00004169 | |
| trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
| CHEBI:45892 | |
| OC(=O)C(F)(F)F |
Safety and Handling
P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
RTECSNumber : AJ9625000