missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific™

Catalog No. AC218861000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC218861000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218861000 Supplier Thermo Scientific Chemicals Supplier No. 218861000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 3336-58-1
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C2H5F3NO2
Linear Formula CF3CO2NH4
MDL Number MFCD00012615,MFCD03095537
Quantity 100g
Beilstein 02,II,186
Synonym ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
Solubility Information Solubility in water: 100g/L (25°C)
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
SMILES [NH4+].OC(=O)C(F)(F)F
IUPAC Name azanium;2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 132.06
PubChem CID 86601334
Formula Weight 131.05
Percent Purity 98%
Chemical Name or Material Trifluoroacetic acid, ammonium salt, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-072-4

RUO – Research Use Only