missing translation for 'onlineSavingsMsg'
Learn More

Acide trifluoroacétique, sel d’ammonium, 98%

Catalog No. AC218860250
Change view
Click to view available options
Quantité:
25 g
Emballage:
Bouteille en verre
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
AC218860250 25 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218860250 Supplier Thermo Scientific Chemicals Supplier No. 218860250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS : 3336-58-1 | C2H5F3NO2 | 132,06 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3336-58-1
Formule moléculaire C2H5F3NO2
Poids moléculaire (g/mol) 132.06
Numéro MDL MFCD00012615,MFCD03095537
Clé InChI YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonyme ammonium trifluoroacetate, azanium 2,2,2-trifluoroacetate, ammoniumtrifluoroacetate, ammonium triflouroacetate, cf3coonh4, acmc-1cq68, 2,2,2-trifluoroacetic acid, azamethane, acetic acid, trifluoro-, ammonium salt 8ci 9ci
PubChem CID 86601334
Nom de l’IUPAC azanium; Acide 2,2,2-trifluoroacétique
SOURIRES [NH4+].OC(=O)C(F)(F)F

Specifications

Nom ou substance chimique Trifluoroacetic acid, ammonium salt
Nom Remarque 0.98
Point de fusion 122,0 °C à 126,0 °C
Couleur Blanc
Spectre infrarouge Authentique
Fourchette de pourcentages d’essai 0.98
Emballage Bouteille en verre
Formule linéaire CF3CO2NH4
Quantité 25 g
Beilstein 02,II,186
Renseignements sur la solubilité Solubilité dans l’eau : 100 g/L (25 °C)
Poids de formule 131.05
Pourcentage de pureté 0.98
Forme physique Poudre cristalline ou cristaux
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-072-4

RUO – Usage réservé à la recherche