missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, 99+%, for HPLC
GREENER_CHOICE

Catalog No. AC432291000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432291000 100 mL Glass bottle
AC432295000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432291000 Supplier Thermo Scientific Chemicals Supplier No. 432291000

missing translation for 'validMainframeMsg'

CAS: 76-05-1, C2HF3O2, 114.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name 2,2,2-trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance 0.03 max. at 300 nm, 0.03 max. at 320 nm, 0.04 max. at 290 nm, 0.05 max. at 280 nm, 0.1 max. at 270nm, 1 max. at 260 nm
Melting Point -15°C
Boiling Point 72°C
Density 1.5350 g/mL
Quantity 100 mL
Infrared Spectrum Conforms to Ref
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.3
Residue after Evaporation 5 ppm max.
Assay Percent Range 99+%
Specific Gravity 1.535
Linear Formula CF3CO2H
Vapor Pressure 107 mbar at 25°C
Green Features ACT Ecolabel (Silver)
Beilstein 02,IV,458
Fieser 09,483; 17,340; 16,83; 15,338; 14,322; 12,529; 10,418; 07,388; 06,613; 05,695; 04,530; 03,305; 02,433; 01,1219; 08,503; 11,557
Merck Index 159847
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ether,acetone,ethanol,benzene,,ccl4,hexane,methanol,xylene and hexane
Formula Weight 114.02
Percent Purity 99+%
Grade HPLC
Chemical Name or Material Trifluoroacetic acid, For HPLC
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 200-929-3

RUO – Research Use Only