missing translation for 'onlineSavingsMsg'
Learn More

Trifluoperazine Dihydrochloride 98.0+%, TCI America™

Catalog No. T28495G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28495G 5 g
T284925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28495G Supplier TCI America Supplier No. T28495G
Only null left

Chemical Identifiers

CAS 440-17-5
Molecular Formula C21H26Cl2F3N3S
Molecular Weight (g/mol) 480.415
MDL Number MFCD00012656
InChI Key BXDAOUXDMHXPDI-UHFFFAOYSA-N
Synonym trifluoperazine dihydrochloride, trifluoperazine hydrochloride, terfluzine, stelazine, triphthazine, eskazinyl, jatroneural, tryptazine, trazine, fluoperazine
PubChem CID 66064
ChEBI CHEBI:9710
IUPAC Name 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride
SMILES CN1CCN(CC1)CCCN2C3=CC=CC=C3SC4=C2C=C(C=C4)C(F)(F)F.Cl.Cl

Specifications

Melting Point 293°C
Color White-Yellow
Quantity 5 g
Formula Weight 480.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Trifluoperazine Dihydrochloride
RTECSNumber SP1750000
TSCA Yes
Recommended Storage Freezer