missing translation for 'onlineSavingsMsg'
Learn More

Triethylvinylsilane 96.0+%, TCI America™

Catalog No. T17035ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17035ML 5 mL
T170325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17035ML Supplier TCI America Supplier No. T17035ML

Chemical Identifiers

CAS 1112-54-5
Molecular Formula C8H18Si
Molecular Weight (g/mol) 142.32
MDL Number MFCD00041345
InChI Key HBWGDHDXAMFADB-UHFFFAOYSA-N
Synonym vinyltriethylsilane, triethylvinylsilane, triethyl vinyl silane, vinyl triethyl silane, ethenyl-triethylsilane, triethylsilyl ethylene, ethenyl triethyl silane, pubchem13130, triethyl vinyl silane #
PubChem CID 559195
IUPAC Name ethenyltriethylsilane
SMILES CC[Si](CC)(CC)C=C

Specifications

Color Yellow
Boiling Point 147°C
Quantity 5 mL
UN Number 1993
Formula Weight 142.32
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylvinylsilane