missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. T16895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16895G 5 g
T168925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16895G Supplier TCI America Supplier No. T16895G

Chemical Identifiers

CAS 79271-56-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000407
InChI Key STMPXDBGVJZCEX-UHFFFAOYSA-N
Synonym triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
PubChem CID 2733308
IUPAC Name triethylsilyl trifluoromethanesulfonate
SMILES CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 79°C
Quantity 5 g
UN Number 1760
Formula Weight 264.33
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Triethylsilyl Trifluoromethanesulfonate