missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl trifluoromethanesulfonate, 99%

Catalog No. AC312700500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC312700500 50 g Glass bottle
AC312700100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC312700500 Supplier Thermo Scientific Chemicals Supplier No. 312700500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79271-56-0 | C7H15F3O3SSi | 264.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Silylation reagent

Chemical Identifiers

CAS 79271-56-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000407
InChI Key STMPXDBGVJZCEX-UHFFFAOYSA-N
Synonym triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
PubChem CID 2733308
IUPAC Name triethylsilyl trifluoromethanesulfonate
SMILES CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F

Specifications

Color Colorless to Brown
Density 1.1600g/mL
Boiling Point 85.0°C to 86.0°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3890 to 1.3920
Linear Formula CF3SO3Si(C2H5)3
Quantity 50 g
Specific Gravity 1.16
Solubility Information Solubility in water: soluble
Formula Weight 264.34
Percent Purity 99%
Physical Form Fuming Liquid
Chemical Name or Material Triethylsilyl trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 279-124-4

RUO – Research Use Only