missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Triethylenetetramine

Numéro de catalogue. 9046050ML
Encompass
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
9046050ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 9046050ML Fournisseur Sigma Aldrich Code fournisseur 9046050ML*
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethylenetetramine

Spécifications

CAS 112-24-3
Melting Point 12°C (lit.)
Density 0.982 g/mL (at 25°C (literature))
Boiling Point 266°C to 267°C (lit.)
Molecular Formula C6H18N4
Refractive Index n20/D 1.496 (literature)
Linear Formula NH2CH2CH2(NHCH2CH2)2 NH2
MDL Number MFCD00008169
Quantity 50 mL
Synonym TETA; DEH 24; EPH 925
Molecular Weight (g/mol) 146.23
Percent Purity ≥97.0% (T)
Afficher plus de résultats Afficher moins de résultats