missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethacrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. T0948500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0948500ML 500 mL
T094825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0948500ML Supplier TCI America Supplier No. T0948500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-16-0
Molecular Formula C14H22O6
Molecular Weight (g/mol) 286.324
MDL Number MFCD00008591
InChI Key HWSSEYVMGDIFMH-UHFFFAOYSA-N
Synonym triethylene glycol dimethacrylate, tedma, nk ester 3g, triethylene dimethacrylate, tgm 3pc, tgm 3s, tgm 3, tgm 35, ethane-1,2-diylbis oxy bis ethane-2,1-diyl bis 2-methylacrylate, polyester tgm 3
PubChem CID 7979
IUPAC Name 2-[2-[2-(2-methylprop-2-enoyloxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOC(=O)C(=C)C

Specifications

Color Yellow
Boiling Point 155°C
Quantity 500 mL
Formula Weight 286.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Dimethacrylate (stabilized with MEHQ)
EINECSNumber (7)-1009&(7)-1438
RTECSNumber OZ4100000
TSCA Yes