missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethacrylate, contains MEHQ as inhibitor

Catalog No. AC466272500
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466272500 250 mL
AC466270010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466272500 Supplier Thermo Scientific Chemicals Supplier No. 466272500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-16-0 | C14H22O6 | 286.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-16-0
Molecular Formula C14H22O6
Molecular Weight (g/mol) 286.32
InChI Key HWSSEYVMGDIFMH-UHFFFAOYSA-N
IUPAC Name 2-(2-{2-[(2-methylprop-2-enoyl)oxy]ethoxy}ethoxy)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCOCCOCCOC(=O)C(C)=C

Specifications

Density 1.0920 g/mL
Boiling Point 170.0°C to 172.0°C (5.0 mmHg)
Flash Point 167°C
Quantity 250 mL
Specific Gravity 1.092
Solubility Information Solubility aq. soln.: 3.6 g/L (20°C). Other solubilities: miscible with most organic solvents
Viscosity 11 mPa.s (25°C)
Formula Weight 286.32
Chemical Name or Material Triethylene glycol dimethacrylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement May cause an allergic skin reaction.

Precautionary Statement GHS P Statement Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

If skin irritation or rash occurs: Get medical advice/attention.

EINECSNumber 203-652-6

RUO – Research Use Only