missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triéthylamine 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -115°C |
| Color | Incolore |
| Boiling Point | 89°C |
| Quantity | 25 mL |
| UN Number | 1296 |
| Formula Weight | 101.19 |
| Percent Purity | ≥99.0% (GC,T) |
| Physical Form | Liquide |
| Chemical Name or Material | Triethylamine |
Sécurité et manipulation
| EINECSNumber | (2)-0141 |
| RTECSNumber | YE0175000 |
| TSCA | Yes |