missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog No. AAL00339AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL00339AE 100 mL
AAL00339AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL00339AE Supplier Thermo Scientific Chemicals Supplier No. L00339AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 122-52-1
Molecular Formula C6H15O3P
Molecular Weight (g/mol) 166.157
MDL Number MFCD00009084
InChI Key BDZBKCUKTQZUTL-UHFFFAOYSA-N
Synonym phosphorous acid, triethyl ester, triethoxyphosphine, ethyl phosphite, eto 3p, triethylphosphite, phosphorous acid triethyl ester, fosforyn trojetylowy czech, unii-6b2r04s55g, hsdb 895, p oet 3, c2h5o 3p
PubChem CID 31215
IUPAC Name triethyl phosphite
SMILES CCOP(OCC)OCC

Specifications

Melting Point -112°C
Density 0.958
Boiling Point 155°C to 157°C
Flash Point 43°C (109°F)
Odor Unpleasant
Refractive Index 1.413
Quantity 100 mL
UN Number UN2323
Beilstein 956578
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 166.16
Percent Purity 98%
Chemical Name or Material Triethyl phosphite
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H332-H319-H317
Flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIETHYL PHOSPHITE
EINECSNumber 204-552-5
RTECSNumber TH1130000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only