missing translation for 'onlineSavingsMsg'
Learn More

Triethyl phosphate, 99+%

Catalog No. AA4000136
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4000136 500 g
AA4000114 25 g
AA4000122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4000136 Supplier Thermo Scientific Chemicals Supplier No. 04000136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol

Triethyl phosphate is used as a solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 78-40-0
Molecular Formula C6H15O4P
Molecular Weight (g/mol) 182.16
MDL Number MFCD00009077
InChI Key DQWPFSLDHJDLRL-UHFFFAOYSA-N
Synonym triethylphosphate, phosphoric acid, triethyl ester, tris ethyl phosphate, triethoxyphosphine oxide, triethylfosfat, ethyl phosphate eto 3po, triethylfosfat czech, ethyl phosphate van, unii-qih4k96k7j, phosphoric acid triethyl ester
PubChem CID 6535
ChEBI CHEBI:45927
IUPAC Name triethyl phosphate
SMILES CCOP(=O)(OCC)OCC

Specifications

Density 1.072 g/mL
Boiling Point 215°C
Flash Point 115°C (240°F)
Odor Mild
Refractive Index 1.405
Quantity 500 g
Merck Index 14,9674
Solubility Information Soluble in water.
Formula Weight 182.16
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Triethyl phosphate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 201-114-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only