missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Orthobenzoate 97.0+%, TCI America™

Catalog No. T30705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30705G 5 g
T307025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30705G Supplier TCI America Supplier No. T30705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1663-61-2
Molecular Formula C13H20O3
Molecular Weight (g/mol) 224.3
MDL Number MFCD00009222
InChI Key BQFPCTXLBRVFJL-UHFFFAOYSA-N
Synonym triethoxymethyl benzene, triethyl orthobenzoate, ethyl orthobenzoate, benzene, triethoxymethyl, triethylorthobenzoate, orthobenzoic acid, triethyl ester, orthobenzoic acid triethyl ester, triethoxymethyl benzol, triethoxyphenylmethane
PubChem CID 74268
IUPAC Name triethoxymethylbenzene
SMILES CCOC(C1=CC=CC=C1)(OCC)OCC

Specifications

Color Colorless
Boiling Point 240°C
Quantity 5 g
Formula Weight 224.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethyl Orthobenzoate