missing translation for 'onlineSavingsMsg'
Learn More

Triethyl Borate 97.0+%, TCI America™

Catalog No. B0520500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0520500ML 500 mL
B052025ML 25 mL
B0520100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0520500ML Supplier TCI America Supplier No. B0520500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 150-46-9
Molecular Formula C6H15BO3
Molecular Weight (g/mol) 145.993
MDL Number MFCD00009073
InChI Key AJSTXXYNEIHPMD-UHFFFAOYSA-N
Synonym triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester
PubChem CID 9009
ChEBI CHEBI:38916
IUPAC Name triethyl borate
SMILES B(OCC)(OCC)OCC

Specifications

Melting Point -85°C
Color Yellow
Boiling Point 118°C
Quantity 500 mL
UN Number 1176
Formula Weight 145.99
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Triethyl Borate
EINECSNumber (2)-2101
RTECSNumber ED5075000
TSCA Yes