missing translation for 'onlineSavingsMsg'
Learn More

Triethyl borate, 97%

Catalog No. AAA18728AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA18728AP 500 mL
AAA18728AE 100 mL
AAA187280F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA18728AP Supplier Thermo Scientific Chemicals Supplier No. A18728AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 150-46-9 | C6H15BO3 | 145.993 g/mol

Triethyl borate is used as a solvent in resins, paints, varnishes, adhesives and sealant chemicals. It is used as a catalyst of synthetic waxes and resins. It is an active component of flame retardants in the textile industry and in welding fluxes. It is used to prepare m-tolyl-acetic acid ethyl ester by reacting with 1-bromomethyl-3-methyl-benzene and carbon monoxide using as1,5-hexadiene?dium chloride as a catalyst. Further, it is used in pyrotechnics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl borate is used as a solvent in resins, paints, varnishes, adhesives and sealant chemicals. It is used as a catalyst of synthetic waxes and resins. It is an active component of flame retardants in the textile industry and in welding fluxes. It is used to prepare m-tolyl-acetic acid ethyl ester by reacting with 1-bromomethyl-3-methyl-benzene and carbon monoxide using as1,5-hexadieneρdium chloride as a catalyst. Further, it is used in pyrotechnics.

Solubility
Miscible with alcohol and ether.

Notes
Moisture sensitive. Incompatible with water, strong oxidizing agents, mineral acids and strong acids.

Chemical Identifiers

CAS 150-46-9
Molecular Formula C6H15BO3
Molecular Weight (g/mol) 145.993
MDL Number MFCD00009073
InChI Key AJSTXXYNEIHPMD-UHFFFAOYSA-N
Synonym triethoxyborane, boron ethoxide, boron triethoxide, triethoxyboron, boric acid, triethyl ester, boric acid triethyl ester, borane, triethoxy, triethylborate, ethyl borate eto 3b, boric acid h3bo3 , triethyl ester
PubChem CID 9009
ChEBI CHEBI:38916
IUPAC Name triethyl borate
SMILES B(OCC)(OCC)OCC

Specifications

Melting Point -85°C
Density 0.858
Boiling Point 117°C to 118°C
Flash Point 11°C (51°F)
Odor Mild
Refractive Index 1.374
Linear Formula (CH3CH2O)3B
Quantity 500 mL
UN Number UN1176
Beilstein 1699052
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohol and ether.
Formula Weight 145.99
Percent Purity 97%
Chemical Name or Material Triethyl borate
Show More Show Less
Hazard Category H225-H336
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHYL BORATE
EINECSNumber 205-760-9
RTECSNumber ED5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only