missing translation for 'onlineSavingsMsg'
Learn More

Triethyl 2-phosphonobutyrate, 97%

Catalog No. AAL0932906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0932906 5 g
AAL0932914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0932906 Supplier Thermo Scientific Chemicals Supplier No. L0932906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 17145-91-4 | C10H21O5P | 252.247 g/mol

Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer?s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant used for preparation of furospinosulin-1 and analogs as hypoxia-selective antitumor agents, Aminoquinolines as beta-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitors and potential anti-Alzheimer′s drugs, phenylpropanoic acid peroxisome proliferator-activated receptor (PPAR) α-selective agonists as nonalcoholic steatohepatitis (NASH)-preventive agents, PPAR agonists with phenethylphenylphthalimide skeleton derived from thalidomide-related LXR antagonists, notch-sparing γ-secretase inhibitors derived from PPAR agonist library and plakotenin via Diels-Alder reaction, as a potential anticancer agent, naturally found in Okinawan sponge of the genus Plakortis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 17145-91-4
Molecular Formula C10H21O5P
Molecular Weight (g/mol) 252.247
MDL Number MFCD00041347
InChI Key GYUCVQSNZFRDRL-UHFFFAOYSA-N
Synonym triethyl 2-phosphonobutyrate, ethyl 2-diethoxyphosphoryl butanoate, ethyl 2-diethylphosphono butyrate, butanoic acid, 2-diethoxyphosphinyl-, ethyl ester, triethyl 2-phosphonobutanoate, .alpha.-diethylphosphono butanoic acid, ethyl ester, butyric acid, 2-phosphono-, triethyl ester, butyric acid, triethyl ester, triethyl 2-phosphonobu-tyrate, triethyl-2-phosphono butyrate
PubChem CID 229053
IUPAC Name ethyl 2-diethoxyphosphorylbutanoate
SMILES CCC(C(=O)OCC)P(=O)(OCC)OCC

Specifications

Density 1.064
Boiling Point 105°C to 106°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.433
Quantity 5 g
Beilstein 1789280
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 252.25
Percent Purity 97%
Chemical Name or Material Triethyl 2-phosphonobutyrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only