missing translation for 'onlineSavingsMsg'
Learn More

Triethoxy-n-octylsilane 97.0+%, TCI America™

Catalog No. O0171500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0171500ML 500 mL
O017125ML 25 mL
O0171100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O0171500ML Supplier TCI America Supplier No. O0171500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Color Colorless
Boiling Point 106°C
Quantity 500 mL
Formula Weight 276.49
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethoxy-n-octylsilane
EINECSNumber (2)-3784
TSCA Yes