missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, Technical, 85%, Spectrum™ Chemical

Catalog No. 18614543
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18614543 500 mL Drum
18614545 4 L Drum
18614544 20 L Drum
18614542 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18614543 Supplier Spectrum Chemical Mfg Cor Supplier No. T2561500MLGL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 102-71-6

Specifications

CAS Max % 1
Boiling Point 335°C
Packaging Drum
Quantity 500 mL
Percent Purity 0.85
Grade Technical
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material Triethanolamine

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.